4-(6-Amino-3-pyridyl)-1-boc-piperazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Boc-4-(6-amino-3-pyridyl)piperazine
, tert-Butyl 4-(6-Amino-3-pyridyl)piperazine-1-carboxylate
, 4-(6-Amino-3-pyridyl)piperazine-1-carboxylic acid tert-Butyl ester
, tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate
, 1-tert-Butoxycarbonyl-4-(6-amino-3-pyridyl)piperazine
H54315-03
H54315-MD
H54315-06
AAH54315-06EA
785.99
USD
AAH54315-06
AAH54315-03
AAH54315-MD
4-(6-Amino-3-pyridyl)-1-boc-piperazine 97%
4-(6-Amino-3-pyridyl)-1-boc-piperazine
Formula:
C₁₄H₂₂N₂O₄ MW: 278.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11594962 CAS Number: 571188-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...