1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole 97%

Warning

H54303-MD H54303-06 H54303-03
AAH54303-06EA 873.89 USD
AAH54303-06 AAH54303-MD AAH54303-03
1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole 97%
1-Boc-4,5-dicyano-2-(3-pyridyl)imidazole
Formula: C₁₅H₁₃N₅O₂
Storage Temperature: Ambient
MDL Number: MFCD20265231
CAS Number: 1347815-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR