3-Amino-5-bromo-2-methoxypyridine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-2-methoxypyridin-3-amine
H54299-06
H54299-MD
H54299-03
AAH54299-06EA
905.99
USD
AAH54299-06
AAH54299-MD
AAH54299-03
3-Amino-5-bromo-2-methoxypyridine 96%
3-Amino-5-bromo-2-methoxypyridine
Formula:
C₆H₇BrN₂O MW: 203.04 g/mol Melting Pt: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368887 CAS Number: 884495-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...