5-(3,5-Dichlorophenylcarbamoyl)-2-fluorophenylboronic acid 97%
|
Warning
|
Synonyms:
5-(3,5-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
H54296-MD
H54296-03
AAH54296-MDEA
96.68
USD
AAH54296-MD
AAH54296-03
5-(3,5-Dichlorophenylcarbamoyl)-2-fluorophenylboronic acid 97%
5-(3,5-Dichlorophenylcarbamoyl)-2-fluorophenylboronic acid
Formula:
C₁₃H₉BCl₂FNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD20040167 CAS Number: 1449132-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...