4,5-Dicyano-2-(3-methylphenyl)imidazole 97%

Danger

H54292-03 H54292-06 H54292-MD
AAH54292-MDEA 27.67 USD
AAH54292-MD AAH54292-03 AAH54292-06
4,5-Dicyano-2-(3-methylphenyl)imidazole 97%
4,5-Dicyano-2-(3-methylphenyl)imidazole
Formula: C₁₂H₈N₄
Storage Temperature: Ambient
MDL Number: MFCD20265228
CAS Number: 1347815-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR