1-Boc-4,5-dicyano-2-(4-pyridyl)imidazole 97%
|
Warning
|
H54232-03
H54232-MD
H54232-06
AAH54232-03EA
206.88
USD
AAH54232-03
AAH54232-06
AAH54232-MD
1-Boc-4,5-dicyano-2-(4-pyridyl)imidazole 97%
1-Boc-4,5-dicyano-2-(4-pyridyl)imidazole
Formula:
C₁₅H₁₃N₅O₂ Storage Temperature: Ambient |
MDL Number:
MFCD20265211 CAS Number: 1347815-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...