Isocytosine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Aminopyrimidin-4(1H)-one
, 2-Amino-4(1H)-pyrimidinone
H54228-03
H54228-14
H54228-06
AAH54228-06EA
108.99
USD
AAH54228-06
AAH54228-03
AAH54228-14
Isocytosine 99%
Isocytosine
Formula:
C₄H₅N₃O MW: 111.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00057557 CAS Number: 108-53-2 EINECS: 203-592-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...