2-Fluoro-5-(2-methoxy-4-nitrophenylcarbamoyl)phenylboronic acid 97%
|
Warning
|
Synonyms:
2-Fluoro-5-(2-methoxy-4-nitrophenylcarbamoyl)benzeneboronic acid
H54224-MD
H54224-03
AAH54224-03EA
268.01
USD
AAH54224-03
AAH54224-MD
2-Fluoro-5-(2-methoxy-4-nitrophenylcarbamoyl)phenylboronic acid 97%
2-Fluoro-5-(2-methoxy-4-nitrophenylcarbamoyl)phenylboronic acid
Formula:
C₁₄H₁₂BFN₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD20265209 CAS Number: 1449144-38-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...