5-(2,6-Dimethylphenylcarbamoyl)-2-fluorophenylboronic acid 97%
|
Warning
|
Synonyms:
5-(2,6-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid
H54222-03
H54222-MD
AAH54222-03EA
382.61
USD
AAH54222-03
AAH54222-MD
5-(2,6-Dimethylphenylcarbamoyl)-2-fluorophenylboronic acid 97%
5-(2,6-Dimethylphenylcarbamoyl)-2-fluorophenylboronic acid
Formula:
C₁₅H₁₅BFNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD20040158 CAS Number: 1449142-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...