2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid 97%
|
Warning
|
Synonyms:
2-Fluoro-5-(4-morpholinylcarbonyl)benzeneboronic acid
H54213-MD
H54213-03
AAH54213-MDEA
114.33
USD
AAH54213-MD
AAH54213-03
2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid 97%
2-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid
Formula:
C₁₁H₁₃BFNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD11504847 CAS Number: 1072951-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...