3-(Hydroxymethyl)benzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Cyanobenzyl alcohol
H54212-MD
H54212-06
H54212-03
AAH54212-03EA
73.09
USD
AAH54212-03
AAH54212-06
AAH54212-MD
3-(Hydroxymethyl)benzonitrile 97%
3-(Hydroxymethyl)benzonitrile
Formula:
C₈H₇NO MW: 133.15 g/mol Flash Pt: 109 °C(228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01321087 CAS Number: 874-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...