3-Methoxy-5-quinolineboronic acid pinacol ester 96%
Supplier: Thermo Scientific Chemicals
H54211-03
H54211-MD
AAH54211-03EA
455.99
USD
AAH54211-03
AAH54211-MD
3-Methoxy-5-quinolineboronic acid pinacol ester 96%
3-Methoxy-5-quinolineboronic acid pinacol ester
Formula:
C₁₆H₂₀BNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD20527212 CAS Number: 881656-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...