4-Fluoro-3-(2-pyridylcarbamoyl)phenylboronic acid 97%
|
Warning
|
Synonyms:
4-Fluoro-3-(2-pyridylcarbamoyl)benzeneboronic acid
H54210-MD
H54210-03
AAH54210-03EA
268.01
USD
AAH54210-03
AAH54210-MD
4-Fluoro-3-(2-pyridylcarbamoyl)phenylboronic acid 97%
4-Fluoro-3-(2-pyridylcarbamoyl)phenylboronic acid
Formula:
C₁₂H₁₀BFN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD20040156 CAS Number: 1449131-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...