2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester 96%
|
Warning
|
H54203-03
H54203-MD
AAH54203-MDEA
136.53
USD
AAH54203-MD
AAH54203-03
2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester 96%
2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester
Formula:
C₂₁H₃₄BN₃O₄ MW: 403.32 g/mol Melting Pt: 132…134 °C Storage Temperature: Ambient |
MDL Number:
MFCD13195039 CAS Number: 1042917-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...