5-(3,4-Dichlorophenylcarbamoyl)-2-fluorophenylboronic acid 97%
|
Warning
|
Synonyms:
5-(3,4-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
H54152-03
H54152-MD
AAH54152-03EA
306.36
USD
AAH54152-03
AAH54152-MD
5-(3,4-Dichlorophenylcarbamoyl)-2-fluorophenylboronic acid 97%
5-(3,4-Dichlorophenylcarbamoyl)-2-fluorophenylboronic acid
Formula:
C₁₃H₉BCl₂FNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD20040143 CAS Number: 1449142-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...