5-(5-Chloro-2-nitrophenylcarbamoyl)-2-fluorophenylboronic acid 97%
|
Warning
|
Synonyms:
5-(5-Chloro-2-nitrophenylcarbamoyl)-2-fluorobenzeneboronic acid
H54121-MD
H54121-03
AAH54121-MDEA
84.12
USD
AAH54121-MD
AAH54121-03
5-(5-Chloro-2-nitrophenylcarbamoyl)-2-fluorophenylboronic acid 97%
5-(5-Chloro-2-nitrophenylcarbamoyl)-2-fluorophenylboronic acid
Formula:
C₁₃H₉BClFN₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD20040135 CAS Number: 1449144-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...