4-(4-Methylpiperazin-1-ylmethyl)benzoic acid 97%
|
Warning
|
Synonyms:
4-((4-Methylpiperazin-1-yl)methyl)benzoic acid
H54120-03
H54120-MD
AAH54120-MDEA
53.72
USD
AAH54120-MD
AAH54120-03
4-(4-Methylpiperazin-1-ylmethyl)benzoic acid 97%
4-(4-Methylpiperazin-1-ylmethyl)benzoic acid
Formula:
C₁₃H₁₈N₂O₂ MW: 234.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01632530 CAS Number: 106261-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...