2-Boc-4'-(Bromomethyl)biphenyl 97%

Supplier: Thermo Scientific Chemicals
Warning

H54100-03 H54100-06 H54100-MD
AAH54100-03EA 129.99 USD
AAH54100-03 AAH54100-06 AAH54100-MD
2-Boc-4'-(Bromomethyl)biphenyl 97%
2-Boc-4'-(Bromomethyl)biphenyl
Formula: C18H19BrO2
Storage Temperature: Ambient
MDL Number: MFCD06657561
CAS Number: 114772-40-6
EINECS: 442-850-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR