5-(2-Chloro-6-nitrophenylcarbamoyl)-3-fluorophenylboronic acid 97%
|
Warning
|
Synonyms:
5-(2-Chloro-6-nitrophenylcarbamoyl)-3-fluorobenzeneboronic acid
H54080-MD
H54080-03
AAH54080-03EA
268.01
USD
AAH54080-03
AAH54080-MD
5-(2-Chloro-6-nitrophenylcarbamoyl)-3-fluorophenylboronic acid 97%
5-(2-Chloro-6-nitrophenylcarbamoyl)-3-fluorophenylboronic acid
Formula:
C₁₃H₉BClFN₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD20126195 CAS Number: 1449133-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...