2-(4-Methoxyphenyl)-4-methyl-5-thiazolecarboxylic acid 97%
|
Warning
|
H54038-MD
H54038-03
H54038-06
AAH54038-03EA
88.42
USD
AAH54038-03
AAH54038-MD
AAH54038-06
2-(4-Methoxyphenyl)-4-methyl-5-thiazolecarboxylic acid 97%
2-(4-Methoxyphenyl)-4-methyl-5-thiazolecarboxylic acid
Formula:
C₁₂H₁₁NO₃S Melting Pt: 236…241 °C Storage Temperature: Ambient |
MDL Number:
MFCD06857928 CAS Number: 54001-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...