O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium hexafluorophosphate

H54035-03 H54035-14 H54035-06
AAH54035-06EA 210.99 USD
AAH54035-06 AAH54035-03 AAH54035-14
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate ≥98%
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Formula: C₉H₁₆F₆N₃O₃P
MW: 359.21 g/mol
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD01863753
CAS Number: 265651-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR