3-(3,5-Dibromophenyl)propionic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(3,5-Dibromophenyl)propanoic acid
H54012-03
H54012-06
AAH54012-06EA
328.99
USD
AAH54012-06
AAH54012-03
3-(3,5-Dibromophenyl)propionic acid 97%
3-(3,5-Dibromophenyl)propionic acid
Formula:
C₉H₈Br₂O₂ MW: 307.97 g/mol Melting Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD11973898 CAS Number: 923977-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...