3-(3,5-Difluorophenyl)propionic acid 97%
|
Danger
|
H54004-14
H54004-06
AAH54004-06EA
89.34
USD
AAH54004-06
AAH54004-14
3-(3,5-Difluorophenyl)propionic acid 97%
3-(3,5-Difluorophenyl)propionic acid
Formula:
C₉H₈F₂O₂ MW: 186.16 g/mol Melting Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD04116054 CAS Number: 84315-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...