5,7-Difluoro-1-indanone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H54002-06
H54002-03
AAH54002-06EA
317.99
USD
AAH54002-06
AAH54002-03
5,7-Difluoro-1-indanone 97%
5,7-Difluoro-1-indanone
Formula:
C₉H₆F₂O MW: 168.14 g/mol Melting Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD09908161 CAS Number: 84315-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...