cis-3-Hexene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(Z)-3-Hexene
H53485-06
H53485-14
AAH53485-06EA
89.59
USD
AAH53485-06
AAH53485-14
cis-3-Hexene 97%
cis-3-Hexene
Formula:
C₆H₁₂ Boiling Pt: 66…68 °C Density: 0.681 Flash Pt: -25 °C(-13 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00085300 CAS Number: 7642-09-3 UN: 3295 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...