Ethynylcyclopentane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H53483-06
H53483-14
H53483-03
AAH53483-06EA
150.99
USD
AAH53483-06
AAH53483-14
AAH53483-03
Ethynylcyclopentane 97%
Ethynylcyclopentane
Formula:
C₇H₁₀ MW: 94.16 g/mol Boiling Pt: 107…109 °C Density: 0.812 Flash Pt: 1 °C(34 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013744 CAS Number: 930-51-8 UN: 3295 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...