1,13-Tetradecadiene 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetradeca-1,13-diene
H53479-03
H53479-14
H53479-06
AAH53479-03EA
53.39
USD
AAH53479-03
AAH53479-14
AAH53479-06
1,13-Tetradecadiene 90+%
1,13-Tetradecadiene
Formula:
C₁₄H₂₆ Boiling Pt: 131 °C/17 mm Density: 0.849 Flash Pt: 103 °C(217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014941 CAS Number: 21964-49-8 EINECS: 244-687-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...