Acetophenone p-toluenesulfonylhydrazone 98%

Supplier: Thermo Scientific Chemicals

H53474-06 H53474-14
AAH53474-06EA 65.59 USD
AAH53474-06 AAH53474-14
Acetophenone p-toluenesulfonylhydrazone 98%
Acetophenone p-toluenesulphonylhydrazone
Formula: C₁₅H₁₆N₂O₂S
Melting Pt: 144…149 °C
Storage Temperature: Ambient
MDL Number: MFCD01140191
CAS Number: 4545-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR