3-(Difluoromethoxy)aniline 97%

Warning

H53468-03 H53468-06 H53468-14
AAH53468-03EA 48.32 USD
AAH53468-03 AAH53468-14 AAH53468-06
3-(Difluoromethoxy)aniline 97%
3-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 201…202 °C
Density: 1.276
Storage Temperature: Ambient
MDL Number: MFCD00236221
CAS Number: 22236-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR