trans-1,2-Dibromocyclohexane 99%
H53456-06
H53456-22
AAH53456-06EA
25.13
USD
AAH53456-06
AAH53456-22
trans-1,2-Dibromocyclohexane 99%
trans-1,2-Dibromocyclohexane
Formula:
C₆H₁₀Br₂ MW: 241.95 g/mol Boiling Pt: 145 °C/100 mm Melting Pt: –6 °C Density: 1.784 Flash Pt: >113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003820 CAS Number: 7429-37-0 EINECS: 231-070-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...