Desoxybenzoin-p-toluenesulfonylhydrazone 97%

H53447-06 H53447-14
AAH53447-06EA 48.99 USD
AAH53447-06 AAH53447-14
Desoxybenzoin-p-toluenesulfonylhydrazone 97%
Desoxybenzoin-p-toluenesulfonylhydrazone
Formula: C₂₁H₂₀N₂O₂S
MW: 364.46 g/mol
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD06411289
CAS Number: 19816-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR