1-(Trimethylsilyl)-1,4-pentadiyne 98%

Supplier: Alfa Aesar
Danger

H53436-06 H53436-03 H53436-14
AAH53436-06EA 95.68 USD InStock
AAH53436-06 AAH53436-03 AAH53436-14
1-(Trimethylsilyl)-1,4-pentadiyne 98%
1-(Trimethylsilyl)-1,4-pentadiyne
Formula: C₈H₁₂Si
MW: 136.27 g/mol
Boiling Pt: 42 °C/25mm
Density: 0,789
Flash Pt: 8 °C (46 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066350
CAS Number: 71789-10-1
EINECS: 000-000-0
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR