N-Butyrophenone-p-toluenesulfonylhydrazone 97%

H53428-06 H53428-14
AAH53428-14EA 169.83 USD
AAH53428-14 AAH53428-06
N-Butyrophenone-p-toluenesulfonylhydrazone 97%
N-Butyrophenone-p-toluenesulfonylhydrazone
Formula: C₁₇H₂₀N₂O₂S
MW: 316.42 g/mol
Melting Pt: 121…125 °C
Storage Temperature: Ambient
MDL Number: MFCD01230699
CAS Number: 41780-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR