3-Iodo-1-phenylpropane 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Iodo-3-phenylpropane

H53420-06 H53420-14 H53420-03
AAH53420-14EA 488.99 USD
AAH53420-14 AAH53420-03 AAH53420-06
3-Iodo-1-phenylpropane 97%
3-Iodo-1-phenylpropane
Formula: C₉H₁₁I
MW: 246.09 g/mol
Density: 1.530
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00019036
CAS Number: 4119-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR