DL-Isoserine 98%
Synonyms:
DL-3-Amino-2-hydroxypropionic Acid
H53412-03
H53412-06
AAH53412-03EA
200.99
USD
AAH53412-03
AAH53412-06
DL-Isoserine 98%
DL-Isoserine
Formula:
C₃H₇NO₃ MW: 105.09 g/mol Melting Pt: 235 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008138 CAS Number: 565-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...