2-Isopropylanisole 98%

Supplier: Thermo Scientific Chemicals

Warning

H53383-14 H53383-22 H53383-06
AAH53383-22EA 252 USD
AAH53383-22 AAH53383-06 AAH53383-14
2-Isopropylanisole 98%
2-Isopropylanisole
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 198…200 °C
Density: 0.936
Flash Pt: 86 °C(187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00798590
CAS Number: 2944-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR