4-(Dimethylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 4-(Dimethylcarbamoyl)benzeneboronic acid(4-(Dimethylcarbamoyl)phenyl)boronic acid

H53363-03 H53363-06
AAH53363-06EA 372.92 USD
AAH53363-06 AAH53363-03
4-(Dimethylcarbamoyl)phenylboronic acid 97%
4-(Dimethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
Melting Pt: 130…136 °C
Storage Temperature: Ambient
MDL Number: MFCD02258943
CAS Number: 405520-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR