4-Methoxy-2,6-dimethylphenylboronic acid 95%

Warning

H53355-03 H53355-MD
AAH53355-MDEA 84.94 USD
AAH53355-MD AAH53355-03
4-Methoxy-2,6-dimethylphenylboronic acid 95%
4-Methoxy-2,6-dimethylphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD03452755
CAS Number: 361543-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR