2-(Ethylthio)phenylboronic acid 97%

Warning

Synonyms: 2-(Ethylthio)benzeneboronic acid

H53350-03 H53350-06
AAH53350-06EA 311.13 USD
AAH53350-06 AAH53350-03
2-(Ethylthio)phenylboronic acid 97%
2-(Ethylthio)phenylboronic acid
Formula: C₈H₁₁BO₂S
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD03095256
CAS Number: 362045-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR