3-Acetamido-2-nitrophenylboronic acid ≥96%

Warning

Synonyms: 3-Acetamido-2-nitrobenzeneboronic acid

H53348-MD
AAH53348-MDEA 382.61 USD
AAH53348-MD
3-Acetamido-2-nitrophenylboronic acid ≥96%
3-Acetamido-2-nitrophenylboronic acid
Formula: C₈H₉BN₂O₅
Melting Pt: 161…162 °C
Storage Temperature: Ambient
MDL Number: MFCD06659828
CAS Number: 78887-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR