4-Chloro-3-(4-morpholinylcarbonyl)phenylboronic acid 98%

Warning

Synonyms: (4-Chloro-3-(morpholine-4-carbonyl)phenyl)boronic acid4-Chloro-3-(4-morpholinylcarbonyl)benzeneboronic acid

H53336-03 H53336-MD
AAH53336-03EA 407.37 USD
AAH53336-03 AAH53336-MD
4-Chloro-3-(4-morpholinylcarbonyl)phenylboronic acid 98%
4-Chloro-3-(4-morpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BClNO₄
MW: 269,49 g/mol
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD07783863
CAS Number: 871332-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR