3-Carboxy-2-methoxyphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Carboxy-2-methoxybenzeneboronic acid3-Borono-2-methoxybenzoic acid

H53323-03 H53323-MD
AAH53323-03EA 428.99 USD
AAH53323-03 AAH53323-MD
3-Carboxy-2-methoxyphenylboronic acid 98%
3-Carboxy-2-methoxyphenylboronic acid
Formula: C₈H₉BO₅
MW: 195.97 g/mol
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD09027237
CAS Number: 913836-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR