3,3'-Carbonylbis(azanediyl)bis(3,1-phenylene)diboronic acid 97%

Warning

Synonyms: 1,3-Bis(3-boronophenyl)urea

H53320-MD H53320-03
AAH53320-03EA 278.61 USD
AAH53320-03 AAH53320-MD
3,3'-Carbonylbis(azanediyl)bis(3,1-phenylene)diboronic acid 97%
3,3'-Carbonylbis(azanediyl)bis(3,1-phenylene)diboronic acid
Formula: C₁₃H₁₄B₂N₂O₅
MW: 299.88 g/mol
Melting Pt: 225…226 °C
Storage Temperature: Ambient
MDL Number: MFCD09027259
CAS Number: 957060-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR