5-Chloro-2-ethoxyphenylboronic acid 97%

Warning

Synonyms: 5-Chloro-2-ethoxybenzeneboronic acid

H53312-03 H53312-06
AAH53312-03EA 102.73 USD
AAH53312-03 AAH53312-06
5-Chloro-2-ethoxyphenylboronic acid 97%
5-Chloro-2-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MW: 200.43 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD03427055
CAS Number: 352534-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR