2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester 95%

Supplier: Thermo Scientific Chemicals

H53309-MD H53309-03
AAH53309-03EA 148.99 USD
AAH53309-03 AAH53309-MD
2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester 95%
2-(Bromomethyl)benzeneboronic acid neopentyl glycol ester
Formula: C₁₂H₁₆BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD04039022
CAS Number: 441011-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR