4-(tert-Butylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 4-(tert-Butylcarbamoyl)benzeneboronic acid(4-(tert-Butylcarbamoyl)phenyl)boronic acid

H53301-03 H53301-06
AAH53301-06EA 438.45 USD
AAH53301-06 AAH53301-03
4-(tert-Butylcarbamoyl)phenylboronic acid 97%
4-(tert-Butylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
Melting Pt: 230…246 °C
Storage Temperature: Ambient
MDL Number: MFCD03788413
CAS Number: 850568-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR