2-Formyl-4-thiopheneboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Formylthiophene-4-boronic acid

H53298-03 H53298-06
AAH53298-03EA 113.99 USD
AAH53298-03 AAH53298-06
2-Formyl-4-thiopheneboronic acid 98%
2-Formyl-4-thiopheneboronic acid
Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Melting Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD03002365
CAS Number: 175592-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR