4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride 95%

Warning

Synonyms: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride

H53296-03 H53296-MD
AAH53296-03EA 250.96 USD
AAH53296-03 AAH53296-MD
4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride 95%
4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid hydrochloride
Formula: C₁₂H₁₈BClN₂O₃
MW: 284.55
Melting Pt: 187…189 °C
Storage Temperature: Ambient
MDL Number: MFCD09027217
CAS Number: 913835-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR