3-Fluoro-4-(methylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: (3-Fluoro-4-(methylcarbamoyl)phenyl)boronic acid3-Fluoro-4-(methylcarbamoyl)benzeneboronic acid

H53289-MD H53289-03
AAH53289-MDEA 76.24 USD
AAH53289-MD AAH53289-03
3-Fluoro-4-(methylcarbamoyl)phenylboronic acid 98%
3-Fluoro-4-(methylcarbamoyl)phenylboronic acid
Formula: C₈H₉BFNO₃
MW: 196.97 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD08436026
CAS Number: 849833-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR