4-(Ethylcarbamoyl)-3-fluorophenylboronic acid 98%

Warning

Synonyms: 4-Ethylcarbamoyl-3-fluorobenzeneboronic acid

H53273-MD H53273-03
AAH53273-03EA 325.23 USD
AAH53273-03 AAH53273-MD
4-(Ethylcarbamoyl)-3-fluorophenylboronic acid 98%
4-(Ethylcarbamoyl)-3-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
MW: 211.00
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD08436070
CAS Number: 874289-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR